ContactUsLinkComponent
Multi-Parameter Refractive Index & Density, @ 15, 20 & 25°C (Nominal RI: 1.5463 @ 20°C), LGC Industrial
Multi-Parameter Refractive Index & Density, @ 15, 20 & 25°C (Nominal RI: 1.5463 @ 20°C), LGC Industrial
Catalog # 77863-203
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Parameter Refractive Index & Density, @ 15, 20 & 25°C (Nominal RI: 1.5463 @ 20°C), LGC Industrial
Catalog # 77863-203
Supplier:  LGC Standards
Supplier Number:  ALK-PS-RVD-03
New Product
CAS Number:  
Specifications
  • Description:
    Multi-Parameter Refractive Index & Density, @ 15, 20 & 25°C (Nominal RI: 1.5463 @ 20°C)
  • Size:
    30 mL