Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Alloys and Metal Ore Standards
Solder, Lead-Free, Ø40x15mm, LGC Industrial
Solder, Lead-Free, Ø40x15mm, LGC Industrial
Solder, Lead-Free, Ø40x15mm, LGC Industrial
Catalog # 77865-284
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Solder, Lead-Free, Ø40x15mm, LGC Industrial
Catalog # 77865-284
Supplier:
LGC Standards
Supplier Number:
MBH-74X CA2 C
New Product
CAS Number:
Specifications
Description:
Solder, Lead-Free, Ø40x15mm
Size:
disc
Specifications
About this item
Product Details
Product Details