Aceclofenac EP Impurity H, TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:1216495-92-9
Specifications
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
tert-Butyl methyl ether ACS
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Chloroform, extra dry over Molecular Sieve 99.9% stabilized, AcroSeal™
Supplier: Thermo Scientific Chemicals
n-Pentane 99+% for HPLC
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Carbon disulfide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
Acetonitrile 99.9%, meets analytical specification of Ph.Eur. for HPLC, far-UV
Supplier: Thermo Scientific Chemicals
Tetrahydrofuran, extra dry 99.85% unstabilized, AcroSeal®
Supplier: Thermo Scientific Chemicals
Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh
Supplier: Thermo Scientific Chemicals
Perchloric acid 70% in water ACS
Supplier: Thermo Scientific Chemicals
Trichloroacetic acid (TCA) 99+% ACS
Supplier: Thermo Scientific Chemicals
tert-Butanol ACS
Supplier: Thermo Scientific Chemicals
MARKER 9.22 ICE280 ICE3 PI
Supplier: 0000014126
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
Tetrachloroethylene 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) ACS
Supplier: Thermo Scientific Chemicals
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
Sulfuric acid 96% in water, extra pure
Supplier: Thermo Scientific Chemicals
Acetonitrile, extra dry 99.9+%, AcroSeal™
Supplier: Thermo Scientific Chemicals