Order Entry
United States
ContactUsLinkComponent
 
Small Sample Viscosity (2.882 cSt @ 40°C), Certified @ 40 & 100°C, LGC Industrial
Catalog # 77863-171
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Small Sample Viscosity (2.882 cSt @ 40°C), Certified @ 40 & 100°C, LGC Industrial
Catalog # 77863-171
Supplier:  LGC Standards
Supplier Number:  ALK-HVS01
New Product
CAS Number:  

Specifications

  • Description:
    Small Sample Viscosity (2.882 cSt @ 40°C), Certified @ 40 & 100°C
  • Size:
    60 mL

Specifications

About this item