Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Alloys and Metal Ore Standards
Stainless Steel, Nitronic 50, S20910, LGC Industrial
Stainless Steel, Nitronic 50, S20910, LGC Industrial
Stainless Steel, Nitronic 50, S20910, LGC Industrial
Catalog # 77865-133
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Stainless Steel, Nitronic 50, S20910, LGC Industrial
Catalog # 77865-133
Supplier:
LGC Standards
Supplier Number:
IARM-17D-G
New Product
CAS Number:
Specifications
Description:
Stainless Steel, Nitronic 50, S20910
Size:
each
Specifications
About this item
Product Details
Product Details