N-phthalyl-L-alanine, TLC Pharmaceutical Standards
New Product
- CAS No.:4192-28-3
Frequently Bought Together
CUBIC-L Tissue-Clearing Reagent, for Animals
: TCI America
tert-Butyl methyl ether for HPLC
: Thermo Scientific Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
: Thermo Scientific Chemicals
Iodic acid 99.6% ACS
: Thermo Scientific Chemicals
Hexane (mixture of isomers) 99%, pure
: Thermo Scientific Chemicals
Acetone 99+%, extra pure
: Thermo Scientific Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
: Thermo Scientific Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) ACS
: Thermo Scientific Chemicals
Tetrachloroethylene 99%, Extra Pure
: Thermo Scientific Chemicals
Tetrahydrofuran 99+% stabilized, extra pure
: Thermo Scientific Chemicals
Methanol 99.9% for analysis
: Thermo Scientific Chemicals
Cyclohexane 99+%, pure
: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
: TCI America
Chloroform 99.9% stabilized, ECD tested for residue analysis, for pesticide residue analysis residue <0.0005%
: Thermo Scientific Chemicals
1,2-Dichloroethane 99+%, pure
: Thermo Scientific Chemicals
Chloroform ≥99.8% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Scientific Chemicals
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal™
: Thermo Scientific Chemicals
Petroleum spirit, 35…60 °C ACS
: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.9% for spectroscopy
: Thermo Scientific Chemicals