Order Entry
United States
ContactUsLinkComponent
Refractive Index (Nominal: 1.5440 @ 25°C), Certified @ 20, 25 & 30°C, LGC Industrial
Refractive Index (Nominal: 1.5440 @ 25°C), Certified @ 20, 25 & 30°C, LGC Industrial
Catalog # 77863-200
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Refractive Index (Nominal: 1.5440 @ 25°C), Certified @ 20, 25 & 30°C, LGC Industrial
Catalog # 77863-200
Supplier:  LGC Standards
Supplier Number:  ALK-PS-RI-09
New Product
CAS Number:  

Specifications

  • Description:
    Refractive Index (Nominal: 1.5440 @ 25°C), Certified @ 20, 25 & 30°C
  • Size:
    10 mL

Specifications

About this item