Amtolmetin Guacil, TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:87344-06-7
Specifications
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
n-Hexane 95+% ACS
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Benzyl alcohol, extra dry 98+%, AcroSeal®
Supplier: Thermo Scientific Chemicals
Chloroform, extra dry over Molecular Sieve 99.9% stabilized, AcroSeal™
Supplier: Thermo Scientific Chemicals
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Scientific Chemicals
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal™
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane 99.8+%, Extra Pure
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Ammonia solution 25% in water, Extra Pure
Supplier: Thermo Scientific Chemicals
Iodic acid 99.6% ACS
Supplier: Thermo Scientific Chemicals
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Scientific Chemicals
Sulfuric acid 95% in water ACS
Supplier: Thermo Scientific Chemicals
N-Methyl-2-pyrrolidone (NMP) 98%, pure
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
Ammonia solution 20% in water for analysis
Supplier: Thermo Scientific Chemicals