Atenolol EP Impurity D (Atenolol USP Related Compound D), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
Atenolol EP Impurity D (Atenolol USP Related Compound D), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
About this item
Specifications
- CAS No.:115538-83-5
Specifications
Frequently Bought Together
Chloroform 99+% stabilized for analysis
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Orthoboric acid 99.99% (trace metals basis), extra pure
Supplier: Thermo Scientific Chemicals
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Supplier: Thermo Scientific Chemicals
Water, extra pure, deionized
Supplier: Thermo Scientific Chemicals
Perchloric acid 70% in water ACS
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane 99.8+%, Extra Pure
Supplier: Thermo Scientific Chemicals
Water, gradient grade for HPLC
Supplier: Thermo Scientific Chemicals
Iodic acid 99.6% ACS
Supplier: Thermo Scientific Chemicals
(±)-2-Butanol 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
Chlorobenzene 99+%, pure
Supplier: Thermo Scientific Chemicals
Methanol 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Scientific Chemicals
Chloroacetic acid 99+%
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
MARKER 6.14 MAURICE CIEF PI 210UL
Supplier: 0000014126
2-(Benzyloxy)phenylboronic acid 97%
Supplier: Ambeed
Petroleum spirit, 35…60 °C ACS
Supplier: Thermo Scientific Chemicals
Cyclohexane 99.5% for analysis
Supplier: Thermo Scientific Chemicals
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Scientific Chemicals