Order Entry
United States
ContactUsLinkComponent
Density 25°C (Nominal: 0.8352 g/mL @ 25°C), LGC Industrial
Density 25°C (Nominal: 0.8352 g/mL @ 25°C), LGC Industrial
  77860-299
New Product
 :  
undefined
Density 25°C (Nominal: 0.8352 g/mL @ 25°C), LGC Industrial
  77860-299
 :  ALK-DEN25-04
New Product
 :  

 

  • Description:
    Density 25°C (Nominal: 0.8352 g/mL @ 25°C)
  • Size:
    60 mL