Sulfur-Free Cobalt Concentrate: Co @ approx. 3 wt% in Hydrocarbons, LGC Industrial
Catalog # 77865-520
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Hydrocarbon Oil
- Description:Sulfur-Free Cobalt Concentrate: Co @ approx. 3 wt% in Hydrocarbons
- Size:50 g