Atorvastatin EP Impurity A Calcium Salt (Atorvastatin USP Related Compound A, Defluoro Atorvastatin Calcium Salt), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
Atorvastatin EP Impurity A Calcium Salt (Atorvastatin USP Related Compound A, Defluoro Atorvastatin Calcium Salt), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
About this item
Frequently Bought Together
Chloroform 99+% stabilized for analysis
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
tert-Butyl methyl ether 99%, pure
Supplier: Thermo Scientific Chemicals
Carbon disulfide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Acetone 99+%, extra pure
Supplier: Thermo Scientific Chemicals
Petroleum spirit, 40…60 °C, extra dry (max. 50 ppm H₂O), AcroSeal™
Supplier: Thermo Scientific Chemicals
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Scientific Chemicals
PI MARKER PI 9.46 200UL
Supplier: 0000014126
(R)-Ibuprofen 96%
Supplier: Ambeed
Orthophosphoric acid 85+% for HPLC
Supplier: Thermo Scientific Chemicals
Periodic acid 99+%, extra pure
Supplier: Thermo Scientific Chemicals
Chloroacetic acid 99+% ACS
Supplier: Thermo Scientific Chemicals
Petroleum spirit, 35…60 °C ACS
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
4-Methyl-2-pentanone 98.5+% ACS
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane 99.8+%, Extra Pure
Supplier: Thermo Scientific Chemicals