Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Boron Standards
Boron Standard: B @ 10 µg/mL in H2O, LGC Industrial
Boron Standard: B @ 10 µg/mL in H2O, LGC Industrial
Boron Standard: B @ 10 µg/mL in H2O, LGC Industrial
Catalog # 77863-152
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Boron Standard: B @ 10 µg/mL in H2O, LGC Industrial
Catalog # 77863-152
Supplier:
LGC Standards
Supplier Number:
VHG-LBW-100
New Product
CAS Number:
Specifications
Matrix:
Water
Description:
Boron Standard: B @ 10 µg/mL in H2O
Size:
250 mL
Specifications
About this item
Product Details
Product Details