Order Entry
United States
ContactUsLinkComponent
Density 15°C (Nominal: 0.8811 g/mL @ 15°C), LGC Industrial
Density 15°C (Nominal: 0.8811 g/mL @ 15°C), LGC Industrial
  77860-285
New Product
 :  
undefined
Density 15°C (Nominal: 0.8811 g/mL @ 15°C), LGC Industrial
  77860-285
 :  ALK-DEN15-07
New Product
 :  

 

  • Description:
    Density 15°C (Nominal: 0.8811 g/mL @ 15°C)
  • Size:
    60 mL