Order Entry
United States
ContactUsLinkComponent
Small Sample Viscosity (32.42 cSt @ 40°C), Certified @ 40 & 100°C, LGC Industrial
Small Sample Viscosity (32.42 cSt @ 40°C), Certified @ 40 & 100°C, LGC Industrial
Catalog # 77863-174
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Small Sample Viscosity (32.42 cSt @ 40°C), Certified @ 40 & 100°C, LGC Industrial
Catalog # 77863-174
Supplier:  LGC Standards
Supplier Number:  ALK-HVS04
New Product
CAS Number:  

Specifications

  • Description:
    Small Sample Viscosity (32.42 cSt @ 40°C), Certified @ 40 & 100°C
  • Size:
    60 mL

Specifications

About this item