Atorvastatin EP Impurity H-d5 (Atorvastatin USP Related Compound H-d5, Atorvastatin Lactone-d5), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
Atorvastatin EP Impurity H-d5 (Atorvastatin USP Related Compound H-d5, Atorvastatin Lactone-d5), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
About this item
Frequently Bought Together
2-Propanol 70% in water, Technical Grade
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Dimethyl sulfoxide 99.8+% for HPLC
Supplier: Thermo Scientific Chemicals
Chloroacetic acid 99+%
Supplier: Thermo Scientific Chemicals
Trifluoroacetic acid 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
Sodium hydroxide 98%, pellets, extra pure
Supplier: Thermo Scientific Chemicals
2-(Benzyloxy)phenylboronic acid 97%
Supplier: Ambeed
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
Isobutanol, extra dry 99+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Scientific Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) ACS
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Carbon disulfide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
Hydrochloric acid 36%, Technical Grade
Supplier: Thermo Scientific Chemicals
Sulfuric acid ∼96% in water for analysis
Supplier: Thermo Scientific Chemicals
Perchloric acid 70% in water ACS
Supplier: Thermo Scientific Chemicals
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal™
Supplier: Thermo Scientific Chemicals