Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Viscometry Standards
Type RT60000, Rotational Viscosity, LGC Industrial
Type RT60000, Rotational Viscosity, LGC Industrial
Type RT60000, Rotational Viscosity, LGC Industrial
Catalog # 77863-242
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Type RT60000, Rotational Viscosity, LGC Industrial
Catalog # 77863-242
Supplier:
LGC Standards
Supplier Number:
ALK-VIS-RT60K-600
New Product
CAS Number:
Specifications
Description:
Type RT60000, Rotational Viscosity
Size:
600 mL
Specifications
About this item
Product Details
Product Details