$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Viscometry Standards
Type N120B, Low Temperature Viscosity, LGC Industrial
Type N120B, Low Temperature Viscosity, LGC Industrial
Catalog # 77863-188
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Type N120B, Low Temperature Viscosity, LGC Industrial
Catalog # 77863-188
Supplier:
LGC Standards
Supplier Number:
ALK-N120B
New Product
CAS Number:
Specifications
Description:
Type N120B, Low Temperature Viscosity
Size:
500 mL
Specifications
About this item
Product Details
Product Details