Order Entry
United States
ContactUsLinkComponent
 
Density 25°C (Nominal: 1.0222 g/mL @ 25°C), LGC Industrial
  77860-300
New Product
 :  
undefined
Density 25°C (Nominal: 1.0222 g/mL @ 25°C), LGC Industrial
  77860-300
 :  ALK-DEN25-08
New Product
 :  

 

  • Description:
    Density 25°C (Nominal: 1.0222 g/mL @ 25°C)
  • Size:
    60 mL