Selenium Standard: Se @ 1000 µg/g in Hydrocarbon Oil, LGC Industrial
Catalog # 77865-516
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:75 cSt Mineral Oil
- Description:Selenium Standard: Se @ 1000 µg/g in Hydrocarbon Oil
- Size:100 g