Atenolol EP Impurity E (Atenolol USP Related Compound E), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
Atenolol EP Impurity E (Atenolol USP Related Compound E), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
About this item
Specifications
- CAS No.:141650-31-9
Specifications
Frequently Bought Together
p-Xylene 99% for HPLC
Supplier: Thermo Scientific Chemicals
1-Butanol 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
Potassium hydroxide ca. 85%, pellets ACS
Supplier: Thermo Scientific Chemicals
Diethylene glycol monobutyl ether 99+%
Supplier: Thermo Scientific Chemicals
Trichloroethylene 99.6% stabilized ACS
Supplier: Thermo Scientific Chemicals
Sodium hydroxide 97+%, pellets ACS
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Formamide 99.6% ACS
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Scientific Chemicals
Sulfuric acid 95% in water ACS
Supplier: Thermo Scientific Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Scientific Chemicals
Perchloric acid ∼70% in water for analysis
Supplier: Thermo Scientific Chemicals
1-Butanol, extra dry 99+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
tert-Butanol ACS
Supplier: Thermo Scientific Chemicals
1-Propanol 99.5% for analysis
Supplier: Thermo Scientific Chemicals
Acetonitrile, extra dry 99.9+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
1,2-Dichloroethane, extra dry 99.8%, AcroSeal®
Supplier: Thermo Scientific Chemicals
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Scientific Chemicals