Almonertinib (HS-10296), TLC Pharmaceutical Standards
Supplier: TLC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:1899921-05-1
Specifications
Frequently Bought Together
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Scientific Chemicals
Chloroform 99.9% stabilized, ECD tested for residue analysis, for pesticide residue analysis residue <0.0005%
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
Chloroacetic acid 99+%
Supplier: Thermo Scientific Chemicals
Diethylene glycol monobutyl ether 99+%
Supplier: Thermo Scientific Chemicals
Trichloroethylene 99.6% stabilized ACS
Supplier: Thermo Scientific Chemicals
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Supplier: Thermo Scientific Chemicals
Iodic acid 99.6% ACS
Supplier: Thermo Scientific Chemicals
Hexane (mixture of isomers) 99%, pure
Supplier: Thermo Scientific Chemicals
Sulfuric acid 95% in water ACS
Supplier: Thermo Scientific Chemicals
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Scientific Chemicals
1-PROPANEPHOSPHONIC ANHYDRID 50%ETH 1KG
Supplier: Oakwood Products INC
p-Xylene 99% for HPLC
Supplier: Thermo Scientific Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) ACS
Supplier: Thermo Scientific Chemicals
2-(Benzyloxy)phenylboronic acid 97%
Supplier: Ambeed