$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Anionic Standards
Multi-Element Anioinic Standards
100ug/mL F-, Cl-, SO4(-2) , LGC Industrial
100ug/mL F-, Cl-, SO4(-2) , LGC Industrial
100ug/mL F-, Cl-, SO4(-2) , LGC Industrial
Catalog # 77865-510
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
100ug/mL F-, Cl-, SO4(-2) , LGC Industrial
Catalog # 77865-510
Supplier:
LGC Standards
Supplier Number:
VHG-ICM2-500
New Product
CAS Number:
Specifications
Matrix:
Water
Description:
100ug/mL F-, Cl-, SO4(-2)
Size:
500 mL
Specifications
About this item
Product Details
Product Details