Anti-TAOK3 Rabbit Polyclonal Antibody
Catalog # 77847-984
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TAOK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q9H2K8
- Antigen Synonyms:CTCL tumor antigen HD CL 09|CTCL-associated antigen HD-CL-09|Cutaneous T cell lymphoma tumor antigen HD CL 09|Cutaneous T-cell lymphoma-associated antigen HD-CL-09|Dendritic cell derived protein kinase|Dendritic cell-derived protein kinase|DKFZp666H245|DPK|FLJ31808|hKFC A|hKFC-A|JIK|JNK/SAPK inhibitory kinase|JNK/SAPK-inhibitory kinase|Jun kinase inhibitory kinase|Jun kinase-inhibitory kinase|KDS|Kinase from chicken homolog A|MAP3K18|Serine kinase|Serine/threonine-protein kinase TAO3|STE20 like kinase|TAO kinase 3|TAOK3_HUMAN|Thousand and one amino acid protein 3
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human TAOK3
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: TAOK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human