Anti-IFIT5 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-850
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IFIT5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13325
- Antigen Synonyms:FLJ53857|FLJ92678|IFIT-5|IFIT5_HUMAN|Interferon induced protein with tetratricopeptide repeats 5|Interferon-induced protein with tetratricopeptide repeats 5|Retinoic acid and interferon inducible 58 kDa protein|Retinoic acid and interferon inducible protein (58kD)|Retinoic acid- and interferon-inducible 58 kDa protein|RI58
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Interferon-induced protein with tetratricopeptide repeats 5 protein (107-320AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: IFIT5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human