Anti-ACOT9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACOT9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-519
- 77846-520
- Antigen Symbol:
- ACOT9
- ACOT9
- Antigen Synonyms:
- ACOT9_HUMAN|Acyl CoA thioester hydrolase 9|Acyl CoA thioesterase 9|Acyl coenzyme A thioesterase 9|Acyl-CoA thioester hydrolase 9|Acyl-CoA thioesterase 9|Acyl-coenzyme A thioesterase 9|mitochondrial|CGI 16
- ACOT9_HUMAN|Acyl CoA thioester hydrolase 9|Acyl CoA thioesterase 9|Acyl coenzyme A thioesterase 9|Acyl-CoA thioester hydrolase 9|Acyl-CoA thioesterase 9|Acyl-coenzyme A thioesterase 9|mitochondrial|CGI 16
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y305
- Q9Y305
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Acyl-coenzyme A thioesterase 9, mitochondrial protein (1-212AA)
- Recombinant Human Acyl-coenzyme A thioesterase 9, mitochondrial protein (1-212AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together