Anti-C4BPA Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-117
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:C4BPA
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P04003
- Antigen Synonyms:C4b-binding protein alpha chain|C4bp|C4BPA_HUMAN|Complement component 4 binding protein alpha|Complement component 4 binding protein|Proline-rich protein|PrP|RP11-164O23.4
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human C4b-binding protein alpha chain protein (49-597AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: C4BPA
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human