Anti-AKAP7 Rabbit Recombinant Polyclonal Antibody
77850-959
New Product
- Antigen Symbol:AKAP7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O43687
- Antigen Synonyms:A kinase (PRKA) anchor protein 7|A kinase anchor protein 18 kD|A kinase anchor protein 7|A kinase anchor protein 7 isoform alpha|A kinase anchor protein 7 isoform beta|A kinase anchor protein 7 isoform gamma|A kinase anchor protein 7 isoforms alpha and beta|A kinase anchor protein 9 kDa|A kinase anchoring protein 18|A-kinase anchor protein 18 kDa|A-kinase anchor protein 7 isoforms alpha and beta|AKA7A_HUMAN|AKAP 18|AKAP 7|AKAP-7 isoforms alpha and beta|AKAP15|AKAP18|HGNC377|OTTHUMP00000040266|Protein kinase A anchoring protein 7|Protein kinase A anchoring protein 7 isoform alpha|Protein kinase A anchoring protein 7 isoform beta|Protein kinase A anchoring protein 7 isoform gamma|Protein kinase A anchoring protein 7 isoforms alpha/beta|Protein kinase A-anchoring protein 7 isoforms alpha/beta
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human A-kinase anchor protein 7 isoforms alpha and beta protein (1-81AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: AKAP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human