Anti-HSF1 HEK293F cells Recombinant Monoclonal Antibody [clone: 2A7]
77850-271
New Product
- Antigen Symbol:HSF1
- Clonality:Recombinant Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q00613
- Antigen Synonyms:Heat shock factor protein 1|HSF 1|Heat shock transcription factor 1|HSF1
- Modification:phospho-Ser326
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from human Phospho-HSF1 (S326)
- Purification:Affinity purified
- Packaging:Vial
Type:
Antigen: HSF1
Clonality: Recombinant Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human, Mouse, Rat