Order Entry
United States
ContactUsLinkComponent
 
Anti-AKR7A2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-988
Supplier:  AFG Bioscience
New Product
undefined
Anti-AKR7A2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-988
Supplier:  AFG Bioscience
Supplier Number:  A12324-50UL
New Product

Specifications

  • Antigen Symbol:
    AKR7A2
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µl
  • Form:
    Liquid
  • Gene ID:
    O43488
  • Antigen Synonyms:
    AFAR|AFAR1|AFB1 aldehyde reductase 1|AFB1 AR1|AFB1-AR 1|AFB1AR1|Aflatoxin aldehyde reductase|Aflatoxin B1 aldehyde reductase member 2|Aflatoxin beta1 aldehyde reductase|Aiar|AKR7|Aldo keto reductase family 7|Aldo keto reductase family 7 member A2 aflatoxin aldehyde reductase|Aldo keto reductase family 7 member A2|member A2 (aflatoxin aldehyde reductase)|Aldoketoreductase 7|ARK72_HUMAN|SSA reductase|Succinic semialdehyde reductase
  • Storage Buffer:
    PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Aflatoxin B1 aldehyde reductase member 2 protein (100-359AA)
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human