Anti-SOX17 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: SOX17
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-190
- 77843-191
- Antigen Symbol:
- SOX17
- SOX17
- Antigen Synonyms:
- FLJ22252|SOX17_HUMAN|SRY (sex determining region Y) box 17|SRY box 17|SRY related HMG box transcription factor Transcription factor SOX-17|Transcription factor VUR3
- FLJ22252|SOX17_HUMAN|SRY (sex determining region Y) box 17|SRY box 17|SRY related HMG box transcription factor Transcription factor SOX-17|Transcription factor VUR3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6I2
- Q9H6I2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription factor SOX-17 protein (177-414AA)
- Recombinant Human Transcription factor SOX-17 protein (177-414AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together