Anti-HOOK1 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOOK1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-200
- 77844-199
- Antigen Symbol:
- HOOK1
- HOOK1
- Antigen Synonyms:
- A930033L17Rik|Abnormal spermatozoon head shape|azh|h-hHK1|HK1|HOOK 1|Hook homolog 1 (Drosophila)|HOOK1_HUMAN|MGC10642|OTTHUMP00000010548|OTTMUSP00000008480|Protein Hook homolog 1|RP23-80B16.4
- A930033L17Rik|Abnormal spermatozoon head shape|azh|h-hHK1|HK1|HOOK 1|Hook homolog 1 (Drosophila)|HOOK1_HUMAN|MGC10642|OTTHUMP00000010548|OTTMUSP00000008480|Protein Hook homolog 1|RP23-80B16.4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJC3
- Q9UJC3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein Hook homolog 1 protein (301-600AA)
- Recombinant Human Protein Hook homolog 1 protein (301-600AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together