Anti-DIMT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-821
- 77844-820
- Antigen Symbol:
- DIMT1
- DIMT1
- Antigen Synonyms:
- 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase|18S rRNA dimethylase|DIM1|DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae)|DIM1 dimethyladenosine transferase 1 homolog|DIM1 dimethyladenosine transferase 1 like |DIM1 dimethyladenosine transferase 1-like|Dimethyladenosine transferase|DIMT1|DIMT1_HUMAN|DIMT1L|HSA9761|HUSSY5|N''-adenosyl(rRNA) dimethyltransferase|Probable 18S rRNA (adenine(1779) N(6)/adenine(1780) N(6)) dimethyltransferase|Probable 18S rRNA dimethylase|Probable dimethyladenosine transferase|Probable S adenosylmethionine 6 N'|N' adenosyl(rRNA) dimethyltransferase|S adenosylmethionine 6 N'|S-adenosylmethionine-6-N''
- 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase|18S rRNA dimethylase|DIM1|DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae)|DIM1 dimethyladenosine transferase 1 homolog|DIM1 dimethyladenosine transferase 1 like |DIM1 dimethyladenosine transferase 1-like|Dimethyladenosine transferase|DIMT1|DIMT1_HUMAN|DIMT1L|HSA9761|HUSSY5|N''-adenosyl(rRNA) dimethyltransferase|Probable 18S rRNA (adenine(1779) N(6)/adenine(1780) N(6)) dimethyltransferase|Probable 18S rRNA dimethylase|Probable dimethyladenosine transferase|Probable S adenosylmethionine 6 N'|N' adenosyl(rRNA) dimethyltransferase|S adenosylmethionine 6 N'|S-adenosylmethionine-6-N''
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNQ2
- Q9UNQ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable dimethyladenosine transferase protein (1-313AA)
- Recombinant Human Probable dimethyladenosine transferase protein (1-313AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together