Anti-ERGIC3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERGIC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-558
- 77845-559
- Antigen Symbol:
- ERGIC3
- ERGIC3
- Antigen Synonyms:
- 2310015B14Rik|AV318804|C20orf47|CGI 54|D2Ucla1|dJ477O4.2|DKFZp547A2190|Endoplasmic reticulum Golgi intermediate compartment protein 3|endoplasmic reticulum localized protein ERp43|Endoplasmic reticulum-Golgi intermediate compartment protein 3|ERGI3_HUMAN|ERGIC and golgi 3|ergic3|ERV46|NY BR 84|PRO0989|RP23-220D12.2|RP3-477O4.1|SDBCAG84|Serologically defined breast cancer antigen 84|Serologically defined breast cancer antigen NY BR 84|Serologically defined breast cancer antigen NY-BR-84
- 2310015B14Rik|AV318804|C20orf47|CGI 54|D2Ucla1|dJ477O4.2|DKFZp547A2190|Endoplasmic reticulum Golgi intermediate compartment protein 3|endoplasmic reticulum localized protein ERp43|Endoplasmic reticulum-Golgi intermediate compartment protein 3|ERGI3_HUMAN|ERGIC and golgi 3|ergic3|ERV46|NY BR 84|PRO0989|RP23-220D12.2|RP3-477O4.1|SDBCAG84|Serologically defined breast cancer antigen 84|Serologically defined breast cancer antigen NY BR 84|Serologically defined breast cancer antigen NY-BR-84
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Endoplasmic reticulum-Golgi intermediate compartment protein 3 protein (47-341AA)
- Recombinant Human Endoplasmic reticulum-Golgi intermediate compartment protein 3 protein (47-341AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together