Anti-ACADSB Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77850-611
New Product
- Antigen Symbol:ACADSB
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P45954
- Antigen Synonyms:2 MEBCAD|2 methyl branched chain acyl CoA dehydrogenase|2 methylbutyryl CoA dehydrogenase|2 methylbutyryl coenzyme A dehydrogenase|2-MEBCAD|2-methyl branched chain acyl-CoA dehydrogenase|2-methylbutyryl-CoA dehydrogenase|2-methylbutyryl-coenzyme A dehydrogenase|ACAD7|ACDSB_HUMAN|acyl CoA dehydrogenase|short/branched chain|acyl Coenzyme A dehydrogenase short branched chain|mitochondrial|OTTHUMP00000020685|OTTHUMP00000046795|SBCAD|Short/branched chain specific acyl-CoA dehydrogenase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human short/branched chain specific acyl-CoA dehydrogenase, mitochondrial protein (34-253AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: ACADSB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human