Anti-MAGEA2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-303
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAGEA2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P43356
- Antigen Synonyms:MAGE2|MAGEA2A|MAGEA2B|Melanoma-associated antigen 2|Cancer/testis antigen 1.2|CT1.2|MAGE-2 antigen
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Melanoma-associated antigen 2 protein (1-104AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: MAGEA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human