Anti-TRIM11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TRIM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77849-324
- 77849-323
- Antigen Symbol:
- TRIM11
- TRIM11
- Antigen Synonyms:
- RING type E3 ubiquitin transferase E3 ubiquitin protein ligase TRI11_HUMAN|Tripartite motif-containing protein 11|Protein BIA1|BIA1|E3 ubiquitin-protein ligase RNF92|BIA 1|RING finger protein 92|Trim 11|tripartite motif protein 11|RNF 92|Tripartite motif containing 11|TRIM11
- RING type E3 ubiquitin transferase E3 ubiquitin protein ligase TRI11_HUMAN|Tripartite motif-containing protein 11|Protein BIA1|BIA1|E3 ubiquitin-protein ligase RNF92|BIA 1|RING finger protein 92|Trim 11|tripartite motif protein 11|RNF 92|Tripartite motif containing 11|TRIM11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96F44
- Q96F44
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TRIM11
- Fusion protein of Human TRIM11
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together