Anti-RPS6KA1 (phospho-Ser380) HEK293F cells Recombinant Monoclonal Antibody [clone: 1E11]
Catalog # 77850-382
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RPS6KA1
- Clonality:Recombinant Monoclonal
- Clone:1E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q15418
- Antigen Synonyms:Ribosomal protein S6 kinase alpha-1|S6K-alpha-1|90 kDa ribosomal protein S6 kinase 1|p90-RSK 1|p90RSK1|p90S6K|MAP kinase-activated protein kinase 1a|MAPK-activated protein kinase 1a|MAPKAP kinase 1a|MAPKAPK-1a|Ribosomal S6 kinase 1|RSK-1|RPS6KA1|MAPKAPK1A|RSK1
- Modification:phospho-Ser38
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-RPS6KA1 (S380)
- Purification:Affinity purified
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IP: 1:200 to 1:1000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human