Anti-EXOSC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EXOSC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-152
- 77846-153
- Antigen Symbol:
- EXOSC1
- EXOSC1
- Antigen Synonyms:
- 3'-5' exoribonuclease CSL4 homolog|CGI 108|CGI-108|CSL4|CSL4 exosomal core protein homolog|CSL4, S. cerevisiae, homolog of|Csl4p|EXOS1_HUMAN|Exosomal core protein CSL4|Exosome complex component csl4|Exosome component 1|hCsl4p|Homolog of yeast exosomal core protein CSL4|p13|RP11-452K12.9|SKI4|Ski4p
- 3'-5' exoribonuclease CSL4 homolog|CGI 108|CGI-108|CSL4|CSL4 exosomal core protein homolog|CSL4, S. cerevisiae, homolog of|Csl4p|EXOS1_HUMAN|Exosomal core protein CSL4|Exosome complex component csl4|Exosome component 1|hCsl4p|Homolog of yeast exosomal core protein CSL4|p13|RP11-452K12.9|SKI4|Ski4p
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3B2
- Q9Y3B2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Exosome complex component CSL4 protein (9-195AA)
- Recombinant Human Exosome complex component CSL4 protein (9-195AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together