Anti-CENPU Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: CENPU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-959
- 77849-958
- Antigen Symbol:
- CENPU
- CENPU
- Antigen Synonyms:
- CENP U|CENP-50|CENP-U|CENP50|CENPU_HUMAN|CENPU50|centromere protein of 50 kDa|Centromere protein U|FLJ23468|HHV8 LNAIP1|ICEN24|Interphase centromere complex protein 24|Kaposi Sarcoma Herpesvirus latent nuclear antigen interacting protein 1|KLIP-1|KLIP1|KSHV latent nuclear antigen interacting protein 1|KSHV latent nuclear antigen-interacting protein 1|KSHV LNAIP1|LNAIP 1|LNAIP-1|MLF1-interacting protein|MLF1IP|MLF1IP1|PBIP1|Polo-box-interacting protein 1
- CENP U|CENP-50|CENP-U|CENP50|CENPU_HUMAN|CENPU50|centromere protein of 50 kDa|Centromere protein U|FLJ23468|HHV8 LNAIP1|ICEN24|Interphase centromere complex protein 24|Kaposi Sarcoma Herpesvirus latent nuclear antigen interacting protein 1|KLIP-1|KLIP1|KSHV latent nuclear antigen interacting protein 1|KSHV latent nuclear antigen-interacting protein 1|KSHV LNAIP1|LNAIP 1|LNAIP-1|MLF1-interacting protein|MLF1IP|MLF1IP1|PBIP1|Polo-box-interacting protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q71F23
- Q71F23
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CENPU
- Fusion protein of Human CENPU
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together