Anti-ETS2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: ETS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-059
- 77847-060
- Antigen Symbol:
- ETS2
- ETS2
- Antigen Synonyms:
- C ets 2 protein|ETS2_HUMAN|ETS2IT1|Oncogene ETS 2|Oncogene Protein C-ets-2|v ets avian erythroblastosis virus E2 oncogene homolog 2|V ets avian erythroblastosis virus E26 oncogene homolog 2|V ets erythroblastosis virus E26 oncogene homolog 2
- C ets 2 protein|ETS2_HUMAN|ETS2IT1|Oncogene ETS 2|Oncogene Protein C-ets-2|v ets avian erythroblastosis virus E2 oncogene homolog 2|V ets avian erythroblastosis virus E26 oncogene homolog 2|V ets erythroblastosis virus E26 oncogene homolog 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15036
- P15036
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ETS2
- Fusion protein of Human ETS2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together