Anti-GSTA2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: GSTA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77849-295
- 77849-294
- Antigen Symbol:
- GSTA2
- GSTA2
- Antigen Synonyms:
- GST2|Glutathione S-transferase A2|EC 2.5.1.18|GST HA subunit 2|GST class-alpha member 2|GST-gamma|GSTA2-2|GTH2
- GST2|Glutathione S-transferase A2|EC 2.5.1.18|GST HA subunit 2|GST class-alpha member 2|GST-gamma|GSTA2-2|GTH2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09210
- P09210
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human GSTA2
- Fusion protein of Human GSTA2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together