Anti-NDFIP1 Rabbit Polyclonal Antibody
Catalog # 77847-509
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NDFIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BT67
- Antigen Synonyms:0610010M22Rik|Breast cancer associated protein SGA 1M|Breast cancer-associated protein SGA-1M|MGC10924|N4WBP5|Ndfip1|Nedd4 family interacting protein 1|NEDD4 family-interacting protein 1|NEDD4 WW domain binding protein 5|NEDD4 WW domain-binding protein 5|NFIP1_HUMAN|Putative MAPK activating protein PM13|Putative MAPK-activating protein PM13|Putative NF kappa B activating protein 164|Putative NF-kappa-B-activating protein 164|Putative NFKB and MAPK activating protein|Putative NFKB and MAPK-activating protein
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from C-terminal of Human NDFIP1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: NDFIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat