Anti-ACADVL Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: ACADVL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-265
- 77849-266
- Antigen Symbol:
- ACADVL
- ACADVL
- Antigen Synonyms:
- ACAD 6|ACAD6|ACADV_HUMAN|Acyl CoA dehydrogenase very long chain|Acyl Coenzyme A dehydrogenase very long chain|LCACD|mitochondrial|Very long chain specific acyl CoA dehydrogenase|Very long chain specific acyl CoA dehydrogenase mitochondrial|Very long-chain specific acyl-CoA dehydrogenase|VLCAD
- ACAD 6|ACAD6|ACADV_HUMAN|Acyl CoA dehydrogenase very long chain|Acyl Coenzyme A dehydrogenase very long chain|LCACD|mitochondrial|Very long chain specific acyl CoA dehydrogenase|Very long chain specific acyl CoA dehydrogenase mitochondrial|Very long-chain specific acyl-CoA dehydrogenase|VLCAD
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49748
- P49748
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ACADVL
- Synthetic peptide of Human ACADVL
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together