Anti-WNK2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WNK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-425
- 77845-424
- Antigen Symbol:
- WNK2
- WNK2
- Antigen Synonyms:
- KIAA1760|PRKSDCCAG43|P/OKcl.13|Serine/threonine-protein kinase EC 2.7.11.1|Antigen NY-CO-43|Protein kinase lysine-deficient 2|Protein kinase with no lysine 2|Serologically defined colon cancer antigen 43
- KIAA1760|PRKSDCCAG43|P/OKcl.13|Serine/threonine-protein kinase EC 2.7.11.1|Antigen NY-CO-43|Protein kinase lysine-deficient 2|Protein kinase with no lysine 2|Serologically defined colon cancer antigen 43
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3S1
- Q9Y3S1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase WNK2 protein (1177-1464AA)
- Recombinant Human Serine/threonine-protein kinase WNK2 protein (1177-1464AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together