Anti-USP20 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-866
- 77845-867
- Antigen Symbol:
- USP20
- USP20
- Antigen Synonyms:
- Deubiquitinating enzyme 20|hVDU2|KIAA1003|LSFR 3A|LSFR3A|pVHL interacting deubiquitinating enzyme 2|Ubiquitin carboxyl terminal hydrolase 20|Ubiquitin carboxyl-terminal hydrolase 20|ubiquitin specific peptidase 20|Ubiquitin specific processing protease 20|Ubiquitin specific protease 20|Ubiquitin thioesterase 20|Ubiquitin thiolesterase 20|Ubiquitin-specific-processing protease 20|UBP20_HUMAN|USP 20|USP20|VDU 2|VDU2|VHL interacting deubiquitinating enzyme 2|VHL-interacting deubiquitinating enzyme 2
- Deubiquitinating enzyme 20|hVDU2|KIAA1003|LSFR 3A|LSFR3A|pVHL interacting deubiquitinating enzyme 2|Ubiquitin carboxyl terminal hydrolase 20|Ubiquitin carboxyl-terminal hydrolase 20|ubiquitin specific peptidase 20|Ubiquitin specific processing protease 20|Ubiquitin specific protease 20|Ubiquitin thioesterase 20|Ubiquitin thiolesterase 20|Ubiquitin-specific-processing protease 20|UBP20_HUMAN|USP 20|USP20|VDU 2|VDU2|VHL interacting deubiquitinating enzyme 2|VHL-interacting deubiquitinating enzyme 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2K6
- Q9Y2K6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 20 protein (258-417AA)
- Recombinant Human Ubiquitin carboxyl-terminal hydrolase 20 protein (258-417AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together