Anti-CLIC4 Rabbit Recombinant Polyclonal Antibody
77853-253
New Product
- Antigen Symbol:CLIC4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9Y696
- Antigen Synonyms:Chloride intracellular channel 4|Chloride intracellular channel 4 (mitochondrial)|Chloride intracellular channel 4 like|Chloride intracellular channel protein 4|CLIC4_HUMAN|CLIC4L|DKFZP566G223|FLJ38640|H1|HUH1|Intracellular chloride ion channel protein p64H1|MC3S5|mtCLIC|p64H1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chloride intracellular channel protein 4 protein (1-253AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, IHC:1:20 to 1:200
Type:
Antigen: CLIC4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse