Anti-KCNA10 Rabbit Polyclonal Antibody
77849-747
New Product
- Antigen Symbol:KCNA10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q16322
- Antigen Synonyms:KCNA10|Potassium voltage-gated channel subfamily A member 10|Voltage-gated potassium channel subunit Kv1.8
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 2000, IHC: 1:100 to 200
Type:
Antigen: KCNA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat