Anti-ASPH Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-497
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ASPH
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:Q12797
- Antigen Synonyms:ASPH|BAHAspartyl/asparaginyl beta-hydroxylase|EC 1.14.11.16|Aspartate beta-hydroxylase|ASP beta-hydroxylase|Peptide-aspartate beta-dioxygenase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aspartyl/asparaginyl beta-hydroxylase protein (75-270AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ASPH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human