Anti-VPS28 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: VPS28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77849-908
- 77849-909
- Antigen Symbol:
- VPS28
- VPS28
- Antigen Synonyms:
- ESCRT-I complex subunit H-Vacuolar protein sorting 28|Vacuolar protein sorting 28 (yeast)|Vacuolar protein sorting 28 homolog (S. cerevisiae)|Vacuolar protein sorting 28 homolog|Vacuolar protein sorting-associated protein 28 homolog|VPS 28|VPS28_HUMAN|Yeast class E protein Vps28p homolog
- ESCRT-I complex subunit H-Vacuolar protein sorting 28|Vacuolar protein sorting 28 (yeast)|Vacuolar protein sorting 28 homolog (S. cerevisiae)|Vacuolar protein sorting 28 homolog|Vacuolar protein sorting-associated protein 28 homolog|VPS 28|VPS28_HUMAN|Yeast class E protein Vps28p homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UK41
- Q9UK41
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together